Learn More
Bel Art™ Poxygrid Test Tube Rack
Product Code. p-7214238 Shop All SP Scienceware Products
Click to view available options
Positions:
48
Unit Size:
Each
Product Code. 17715834
Brand: Bel Art™ F187840001
Only null left
Add to basket
Add to basket
Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy. Orange racks can be used for biohazard warning, yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Will withstand a temperature range of -223°C (-369°F) to 121°C (250°F); autoclavable
  • All racks have square holes, three decks, a slotted grid bottom and reinforced edges
Specifications
Product Type Test Tube Rack
Array 6 x 8
Autoclavable Autoclavable
Color Blue
Holds Test Tubes
Material Steel Wire
Positions 48
Shape Rectangular
Shelves 3 Tiers
Stackable Yes, When Empty
Sterility Non-sterile
No. per Pack 1 Ea.
Dimensions (L x W x H) 36.5 x 30.4 x 10.2 cm
Height (Metric) 10.2 cm