Learn More
Brady™ PowerMark™ Vinyl Labelling Tape
Product Code. p-8001657
Click to view available options
Dimensions (LxW):
15.24 m x 101.6 mm
15.24 m x 152.4 mm
15.24 m x 203.20 mm
15.24 m x 254 mm
Color:
Black
Blue
Brown
Gold
Gray
Green
Orange
Purple
Red
White
Yellow
Unit Size:
Each
Product Code. 11552695
Brand: Brady™ 013605
Brady's B-595 vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics. PowerMark™ Vinyl Labelling Tape also excels at sticking to highly textured surfaces.
  • Brady's B-595 vinyl will adhere to difficult low surface energy items, including: PVC piping, blow-molded equipment cases, ABS plastics, powder-coated surfaces and equipment made from recycled plastics.
  • The B-595 material also excels at sticking to highly textured surfaces such as painted cinder block walls, finished uneven wood, textured plastics, rough-pipes and paper-jacketed pipes.
Specifications
Certifications/Compliance RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Description BBP85 Tape - Vinyl 150 mm Black
Adhesive Type Permanent Acrylic, Permanent
Color Black
Finish Gloss
Format Roll
Material Vinyl
Print Technology Thermal Transfer
Product Type Label
For Use With (Application) Component and equipment labelling, Electrical labelling, Facility Identification, Floor marking, Laboratory labelling, Lean and 5S labelling, Pipe and valve marking, Warehouse marking
For Use With (Equipment) BBP85, PowerMark Industrial Sign and Label Printer
Gloss 60° 50 Gardner Units
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Roll/Box
Quantity 1 Ea.
Software Type MarkWare
Temperature Range (English) -40°F to +180°F
Temperature Range (Metric) -40°C to +82°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (LxW) 15.24 m x 152.4 mm
Length (English) 50 ft.
Length (Metric) 15.24 m
Width (English) 6 in.
Width (Metric) 152.4 mm
Alternate Part Number B85-150x15M-595-BK