Learn More
Brady™ PowerMark™ Reflective Vinyl Labeling Tape
Product Code. 11562675
Change view
Click to view available options
Width (Metric):
101.6 mm
177.8 mm
254 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) unitSize
11562675 101.6 mm Each
11592675 177.8 mm Each
11522685 254 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11562675 Supplier Brady™ Supplier No. 013580
The reflective quality of the material makes PowerMark™ Reflective Vinyl Labeling Tape ideal for reading labels in low light conditions (with flashlights).
  • The reflective quality of the material makes it ideal for reading labels in low light conditions (with flashlights).
Specifications
Certifications/Compliance Type 1 (Fed. Spec. L-S-300)
Description BBP85 Tape - 100 mm silver Reflective
Adhesive Type Permanent Acrylic
Color Silver
Finish Gloss
Material Plastic
Print Technology Thermal Transfer
Product Type Label
For Use With (Application) Component and equipment labelling, Facility Identification, General Identification, Safety Identification
For Use With (Equipment) BBP85, PowerMark Industrial Sign and Label Printer
Gloss 100 Gardner Units
Labels per Row 1
Life Span 1 Year
Min. Operating Temperature (Metric) -40°C
No. per Pack 1 Roll/Box
Quantity 1 Ea.
Software Type MarkWare
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Tensile Strength PSTC-31: 20 lb./in. (350 N/100 mm)
Thickness (English) 0.0055 in.
Thickness (Metric) 0.14 mm
Dimensions (LxW) 10.06 m x 101.6 mm
Length (English) 33 ft.
Length (Metric) 10.06 m
Width (English) 4 in.
Width (Metric) 101.6 mm
Alternate Part Number B85-100x10M-584-SV