Learn More
Ansell™ PowerFlex™ 80-658 Series Aramid Fiber Heavy Weight Gloves
Product Code. p-8005333 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11726723 10 12 pairs
12647011 11 12 pairs
12657011 6 12 pairs
11725834 8 12 pairs
11736723 9 12 pairs
15646906 7 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11726723 Supplier Ansell™ Supplier No. 80658100
Only null left
Add to basket
Add to basket
Protects wearer with triple-layer construction comprised of steel, glass fiber and aramid fiber provides extreme resistance to cuts, splinters and burrs.
  • Mechanical protection gloves
  • Exceeds EN388 cut level 5.
  • Extra-long cuff offers extended safety for the forearm.
  • ¾ dipped and liquid repellent, shielding the skin from unpleasant liquid solutions.
  • Good heat contact resistance.
  • EN407 level-2 rating means safe handling of warm objects.
  • Natural rubber crinkle finish provides a firm grip.
  • Long life provides a highly cost-effective solution.
  • Heather green color helps mask dirt.
  • Excellent washability, with no decrease in comfort or fading.
Applications
  • Handling heavy loads, metal edges
  • Waste handling: recycling, sorting (glass, cans, metal pieces)
  • Handling glass sheets, sharp-edged objects
  • Handling metal sheets, cutting of dry, painted or galvanized pieces
TRUSTED_SUSTAINABILITY
Specifications
Back Material Dupont™ Kevlar™ aramid fibers
Certifications/Compliance CE Mark, Standard CATEGORY III EN 388 : X543, EN 407
Cuff Material Dupont™ Kevlar™ aramid fibers
Coating Material Latex
Color Blue
Cut Resistance Rating ANSI A5
Exterior Finish Crinkle
Hand Specific
Latex Free No
Material Cotton
Product Type Heavyweight Gloves
Size 10
Glove Cuff Style Long Knitwrist
Liner Material Natural Rubber Latex
Lining Lined
Palm Type 3/4 Dipped palm
Quantity 72 Pairs
Resistant To Good heat and cut resistance