missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian Protection™ Powercoat™ 953-03 Nitraf Gloves
Description
- Nitrile composition for mechanical and chemical resistance to a large range of products including oils, greases, hydrocarbons, detergents and many solvents.
- Suitable for food preparation that includes greasy products, production of paints, varnishes.
- Flocked-cotton inside for comfort and easy donning and doffing.
- Diamond pattern finish for good grip in greasy and damp environments.
- Great elasticity and dexterity.
- Safety Category III glove.
- Long cuff helps to protect a large part of the forearm.
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY III Irreversible risks. EN 420-2003 Basic standard, EN 388-2003 Mechanical risks, EN 374-2003 Chemical risks, EC Category PPE 3, EC Certificate Number 0075/797/162/02/07/0042, EC Attestation Number 0075/797/162/02/07/0042 EXT 02/02/07 |
| Classification | Dexterity: 5 |
| Color | Green |
| Cut Resistance Rating | 1 |
| Material | Cotton, Nitrile |
| Product Type | Gloves |
| Size | 8, 9, 10, 11 |
| Thickness (Metric) | 0.52 mm |
| For Use With (Application) | Agriculture, Automotive, Aviation, Building, Construction, Catering, Chemical, Food, Industrial Cleaning, Iron, Steel, Maintenance, Offshore, Petrochemical, Printing, Service, Ship Building, Textile, Transportation and Utility industries. |
| Liner Material | Cotton flock |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?