Learn More
Testo™ Power Supply
Product Code. 10702954
Change view
Click to view available options
Amperage:
500 mA with European Adapter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Amperage Unit Size
10702954 500 mA with European Adapter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10702954 Supplier Testo™ Supplier No. 0554 0447
  • ONLINE EXCLUSIVE PRICE
    43.70€
    28.00€ / Pack of 1
    Save 15.70€
    35% Off
Estimated Shipment: 04-12-2026
Specifications
Amperage 500 mA with European Adapter
Frequency 50/60 Hz
For Use With (Equipment) Testo™ 435/556/560/570/635/735 Measuring Instruments