Learn More
Bellingham Stanley™ Power supply 230V 50Hz for RFM300 Bellingham and Stanley
Product Code. 11806125
Product Code. 11806125 Supplier Bellingham Stanley™ Supplier No. 55105
  • ONLINE EXCLUSIVE PRICE
    371.00€
    243.82€ / Pack of 1
    Save 127.18€
    34% Off
Estimated Shipment: 13-11-2026
Power Supply for RFM300 Series Refractometers, 230 V, 50 Hz
  • Brand: Bellingham Stanley
  • Product: Power Supply
  • Voltage: 230 V
  • Current: 50 Hz
  • For Use With: RFM300 Series Refractometers