Learn More
Aalborg POWER SUPPLY 230V/15V F/ DFC
Brand:  Aalborg PS-DFC-230EU-5-A
POWER SUPPLY 230V/15V F/ DFC
Product Code. 11882650
  • ONLINE EXCLUSIVE PRICE
    443.10€
    269.28€ / Each
    Save 173.82€
    39% Off
Only null left
Estimated Shipment: 03-03-2026