Learn More
Fisherbrand™ Power Cord
Description
- Power output connectors
- Designed for low-to medium voltage power supplies
- Diameter: 4mm
Specifications
Specifications
| For Use With (Equipment) | TV100YK and TV400YK 2-D Electrophoresis Gel Equipment |
| Type | Power Cord |
| Includes | Output connectors |
| Length (Metric) | 2 x 1 m |
| Electrical Requirements | 230 V 50 Hz |
| Max. Voltage | 230 V |
| Voltage | 230 V |
| Frequency | 50 Hz |