Learn More
Memmert™ Potential-Free Contact (24V/2A) W/Socket To Namur Ne 28 For Combined Error Message, For Hcp105
Product Code. 16314807 Shop All Memmert Products
Click to view available options
Voltage:
24 V
Unit Size:
Each
Product Code. 16314807
Brand: Memmert™ HCP105H6
Only null left
Add to basket
Add to basket
Includes
Socket 3-Pin
Specifications
For Use With (Equipment) Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108
Type Potential-free Contact
Voltage 24 V
Amperage 2 A