missing translation for 'onlineSavingsMsg'
Learn More

Potassium tungsten oxide, 99.5% (metals basis)

Product Code. 11301569
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301569 100 g 100g
11311569 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301569 Supplier Thermo Scientific Alfa Aesar Supplier No. 014031.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-60-5 | K2O4W | 326.033 g/mol

Potassium tungsten oxide is used in solar energy and water treatment applications. It is used as alkaloids precipitating agent, bronze production and in tungsten refining. It is also used as an analytical reagent. Further, it is added to molten carbonate fuel cell to prevent the corrosion of stainless-steel separator and retards the sintering of the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-60-5
Molecular Formula K2O4W
Molecular Weight (g/mol) 326.033
MDL Number MFCD00049668
InChI Key AAQNGTNRWPXMPB-UHFFFAOYSA-N
Synonym potassium tungstate, unii-t6vrm0ou6v, t6vrm0ou6v, dipotassium dioxido dioxo tungsten, dipotassium wolframate, potassium tungstate vi, tungstate wo42-, dipotassium, t-4, 2k.wo4, potassium tungstate-w
PubChem CID 3084032
IUPAC Name dipotassium;dioxido(dioxo)tungsten
SMILES [O-][W](=O)(=O)[O-].[K+].[K+]

Specifications

Melting Point 921°C
Physical Form Powder
Quantity 100 g
Merck Index 14,7694
Solubility Information Soluble in water.
Formula Weight 326.04
Percent Purity 99.5%
Odor Odorless
Sensitivity Hygroscopic
Density 3.12 g/mL
Assay (metals basis)
Chemical Name or Material Potassium tungsten oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.