Learn More
Potassium tetraoxalate dihydrate, Honeywell Fluka™
Product Code. 15675990 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15675990 100 g 100g
15685990 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15675990 Supplier Honeywell-Fluka Supplier No. 60592100G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 6100-20-5
Molecular Formula C4H3KO8
Molecular Weight (g/mol) 218.16
MDL Number MFCD00150443,MFCD00150443
InChI Key GANDVAJEIJXBQJ-UHFFFAOYSA-M
Synonym potassium trihydrogen dioxalate dihydrate, potassium ion oxalic acid dihydrate hydrogen oxalate, potassium oxalic acid dihydrate hydrogen oxalate
PubChem CID 131698588
IUPAC Name potassium oxalic acid hydrogen oxalate
SMILES [K+].OC(=O)C(O)=O.OC(=O)C([O-])=O
Specifications
Quantity 100 g
Linear Formula KH3(C2O4)2 · 2H2O
UN Number NONH for all modes of transport
Beilstein 3854419
Formula Weight 254.19g/mol
Percent Purity ≥99.5%
Chemical Name or Material Potassium tetraoxalate dihydrate