Learn More
Potassium Perfluoro-1-butanesulfonate, TRC
Product Code. 30007074
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007074 25 g 25g
30003298 5 g 5g
17951235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30007074 Supplier Toronto Research Chemicals Supplier No. TRCP68827025G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29420-49-3
Molecular Formula C4 F9 O3 S . K
Molecular Weight (g/mol) 338.19
Synonym 1-Butanesulfonic acid, 1,1,2,2,3,3,4,4,4-nonafluoro-, potassium salt (1:1), 1-Butanesulfonic acid, 1,1,2,2,3,3,4,4,4-nonafluoro-, potassium salt (8CI,9CI), Bayowet C 4, Bayowet C 4TP-AC2001, Biowet C 4, Chemguard 411, EF 42, Eftop KFBS, F 114, F 114P, FR 2025, FR 2050, FR 2225, Flutel RM 65, IHT-FR 21, K-NONA, KFBS, KPFBS, Megafac F 114, Megafac F 114P, Megafac F 144P, Megafac F 411, Nanofluorobutanesulfonic acid potassium salt, Nonafluoro-1-butanesulfonate potassium salt, Nonafluorobutanesulfonic acid potassium salt, PFBSA, PPBS, PPFBS, Perfluorobutanesulfonic acid potassium salt, Poatssium perfluorobutanesulfonate, Potassium nonaflate, Potassium nonafluoro-1-butanesulfonate, Potassium nonafluorobutanesulfonate, Potassium perfluoro-1-butanesulfonate, Potassium perfluorobutanesulfonate, Potassium perfluorobutylsulfonate, RM 65
IUPAC Name potassium;1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonate
SMILES [K+].[O-]S(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifications
Quantity 25 g
InChI Formula InChI=1S/C4HF9O3S.K/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16;/h(H,14,15,16);/q;+1/p-1
Formula Weight 337.9062
Chemical Name or Material Perfluorobutanesulfonic acid potassium salt