missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Octadecanoate-2,2-d2, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Potassium Octadecanoate-2,2-D2 |
| CAS | 352438-86-9 |
| Molecular Formula | C18 D2 H33 O2 . K |
| Quantity | 0.5 g |
| Synonym | Octadecanoic-2,2-d2 acid, potassium salt (9 CI), Octadecanoic-2,2-d2 acid potassium salt, Stearic-2,2-d2 acid potassium salt, Potassium stearate-2,2-d2, Potassium octadecanoate-2,2-d2, Potassium octadecanoate-2,2-D2 |
| InChI Formula | InChI=1 S/C18H36O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h2-17H2,1H3,(H,19,20);/q;+1/p-1/i17D2; |
| SMILES | [K+].[2 H]C([2 H])(CCCCCCCCCCCCCCCC)C(=O)[O-] |
| IUPAC Name | potassium;2,2-dideuteriooctadecanoate |
| Molecular Weight (g/mol) | 324.58 |
| Formula Weight | 324.24 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction