missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydroxide, ACS Reagent Grade, Ricca Chemical

Product Code. 31749536
Change view
Click to view available options
Quantity:
500 g
3 kg
25 kg
1000 gal.
Packaging:
Fiber Drum
Natural Poly Bottle
White Poly Wide Mouth Bottle
Unit Size:
1000g
25kg
3kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
31749536 500 g White Poly Wide Mouth Bottle 500g
31749718 1000 gal. White Poly Wide Mouth Bottle 1000g
31750643 3 kg Natural Poly Bottle 3kg
31750020 25 kg Fiber Drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31749536 Supplier Ricca Chemical Company Supplier No. RDCP0500500B1
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1310-58-3 , 7732-18-5
Molecular Formula HKO
Molecular Weight (g/mol) 56.11
MDL Number MFCD00003553
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
PubChem CID 14797
ChEBI CHEBI:32035
IUPAC Name potassium hydroxide
SMILES [OH-].[K+]

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Quantity 500 g
Percent Purity ≥85%
Grade ACS
Packaging White Poly Wide Mouth Bottle
Physical Form Deliquescent Pellets
Chemical Name or Material Potassium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.