Learn More
Potassium Hydroxide, 0.1M, Chem-Lab
Product Code. 11923173
Change view
Click to view available options
Quantity:
5 L
Packaging:
EP
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11923173 5 L EP 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11923173 Supplier Chem-Lab Supplier No. CL05.1105.5500
For laboratory use, Titrations
Specifications
CAS 1310-58-3
Molecular Formula HKO
Concentration 5.611gKOH / 1L H2O
MDL Number MFCD00003553
UN Number UN1814
Density 1.01g/mL
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
Quantity 5 L
SMILES [OH-].[K+]
IUPAC Name potassium;hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
ChEBI CHEBI:32035
Formula Weight 56.11g/mol
Normality 0.1N (±0.0002 at 20°C)
Packaging EP
Chemical Name or Material Potassium hydroxide
Show More Show Less
compliance-icons
Product Identifier
  • Potassium hydroxide 0.1 mol/l
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: KOH/H2O