Learn More
Potassium Hexadecanoate-2,2-d2, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | Potassium Hexadecanoate-2,2-D2 |
| CAS | 203805-86-1 |
| Molecular Formula | C16 D2 H29 O2 . K |
| Quantity | 0.5 g |
| Synonym | Hexadecanoic-2,2-d2 acid, potassium salt (9 CI), [2,2-2H2]Hexadecanoic acid potassium salt, Hexadecanoic-2,2-d2 acid potassium salt, Hexadecanoic acid-2,2-d2 potassium, Palmitic-2,2-d2 acid potassium salt, Potassium hexadecanoate-2,2-D2 |
| InChI Formula | InChI=1 S/C16H32O2.K/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18;/h2-15H2,1H3,(H,17,18);/q;+1/p-1/i15D2; |
| SMILES | [K+].[2 H]C([2 H])(CCCCCCCCCCCCCC)C(=O)[O-] |
| IUPAC Name | potassium;2,2-dideuteriohexadecanoate |
| Molecular Weight (g/mol) | 296.527 |
| Formula Weight | 296.209 g/mol |
| Show More |