Learn More
Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters
Product Code. 11713333
Change view
Click to view available options
For Use With (Application):
High-Range Detection
Low-Range Detection
Range (Conductivity):
0 to 3999μS/cm
0.00 to 20.00mS/cm
Range (TDS):
0 to 2000ppm (mg/L)
0.00 to 10.00ppt (g/L)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Range (Conductivity) Range (TDS) unitSize
11713333 Low-Range Detection 0 to 3999μS/cm 0 to 2000ppm (mg/L) Each
11723333 High-Range Detection 0.00 to 20.00mS/cm 0.00 to 10.00ppt (g/L) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11713333 Supplier Hanna Instruments™ Supplier No. HI991300N
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Designed to measure pH, conductivity (EC), total dissolved solids (TDS), and temperature with a single probe. Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters are available for high range and low range measurement.
This multi-parameter meter takes readings simply and quickly, and it fits in the palm of your hand. From setup to calibration all operations are performed with just two buttons. It features a clear, extra-large LCD display and easy-access rotary knobs to effect calibration changes. A single pre-amplified probe is used for all parameters.
  • Waterproof and rated for IP 67 conditions
  • Easy to clean and maintain
  • Multi-level LCD display
  • On-screen tutorial messages for calibration and setup
  • Automatic Temperature Compensation
  • Automatic one or two-point pH calibration
  • BEPS: Alerts the user in the event that low battery power could adversely affect the readings
  • Battery % displayed on startup
  • Compact, heavy duty, and waterproof
  • Preprogrammed buffer recognition values, TDS factors and temperature coefficients
Specifications
Includes HI 1288 probe, batteries, 1m cable, rugged carrying case and instructions
Accuracy (Temperature) ±0.5°C/±1°F
Accuracy (pH) ±0.01
For Use With (Application) Low-Range Detection
Calibration Automatic
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Resolution (mV) ±1999mV
Temperature Range (Metric) 0°C to 60.0°C
Temperature Range (English) 32.0°F to 140.0°F
Resolution (Temperature) 0.1°C/F
Display Type LCD
Certifications/Compliance IP 67
Dimensions (L x W x H) 150 x 80 x 36 mm (5.9 x 3.1 x 1.4 in.)
Weight (Metric) 205 g
Weight (English) 7.2 oz.
Warranty 2 Years (6 Months Probe)
Keypad Two-button
Accuracy (Conductivity) ±2% F.S.
Material PVC
Probe Type HI1288 PVC body, pre-amplified multiparameter probe with internal temperature sensor, DIN connector and 1m cable
Range (Conductivity) 0 to 3999μS/cm
Range (TDS) 0 to 2000ppm (mg/L)
Resolution TDS Conversion Factor: 0.45 to 1.00
Resolution (Conductivity) 1μS/cm
Resolution (TDS) 1ppm (mg/L)
Detection Range 0 to 14 pH
Temperature Compensation pH: automatic; EC/TDS: automatic with β adjustable from 0.0 to 2.4%/°C
Battery Type 1.5V (3) AAA, Auto-Off after 8 min. non-use
Type Portable pH/EC/TDS/Temperature Meter
Battery Life 500 hr.
Accuracy (TDS) ±2% full scale