Learn More
Extech Portable Indoor Air Quality CO2 Meter
Product Code. 12200419 Shop All Cole-Parmer Products
Change view
Click to view available options
Measurement Range:
0 to 5000 ppm (CO2), 0.0 to 99.9% (RH)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Unit Size
12200419 0 to 5000 ppm (CO2), 0.0 to 99.9% (RH) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12200419 Supplier Extech Supplier No. 8197390
  • ONLINE EXCLUSIVE PRICE
    745.00€
    556.38€ / Pack of 1
    Save 188.62€
    25% Off
Estimated Shipment: 21-12-2026
Use the Portable Indoor Air Quality CO2 Meter with built-in RS-232 interface for capturing readings on your PC.
  • Check air quality in schools, office buildings, greenhouses, and anywhere that high levels of carbon dioxide are generated.
  • Features a maintenance-free NDIR (nondispersive infrared) CO2 sensor, user-programmable audible alarm, backlit LCD, and RS-232 interface.
  • Meter calculates statistical weighted averages of TWA (8 hour time-weighted average) and STEL (15 minutes short term exposure limit).
Specifications
Detectable Analytes Carbon Dioxide
Product Type Portable Indoor Air Quality CO2 Meter
Temperature (Metric) Operating -10°C to +60°C
Alarms Audible
Connections RS-232
Display Type Backlit LED
Measurement Range 0 to 5000 ppm (CO2), 0.0 to 99.9% (RH)
Outputs RS-232
Temperature (English) Operating 14°F to 140°F