Learn More
Esculape Laboratoire™ Eyewash Bottles for First Aid
Product Code. 11759522
Click to view available options
Volume (Metric):
200 mL
500 mL
Composition:
Sterile Isotonic Solution
Sterile Ophthalmic Solution
Includes:
200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution
Cap
Cap, Storage, Carrying Case
Quantity:
10 Pk.
12 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 12
Product Code. 11759522
Brand: Esculape Laboratoire™ 8205260
Use Esculape™ Eyewash Bottles for First Aid has a carrying case with a belt loop for portabilitiy.
  • Storage case
  • Contain sterile isotonic solution
  • Ergonomic
Specifications
Product Type Eyewash Bottle
Composition Sterile Ophthalmic Solution
For Use With (Application) Portable Eyewash
Includes 200 mL Blue Neutral pH Solution, 50 0mL Bottle Green Sterile Ophthalmic Solution
Quantity Ea.
Sterility Sterile
Volume (Metric) 200 mL