Learn More
Hanna Instruments™ Analog pH/mV/°C Meter with Electrodes
Product Code. 11703639
Change view
Click to view available options
Dimensions (L x W x H):
145 x 80 x 36 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
11703639 145 x 80 x 36 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11703639 Supplier Hanna Instruments™ Supplier No. HI83141N
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to be accurate, reliable and easy to use. Hanna Instruments™ Analog pH/mV/°C Meter with Electrodes is a portable multiparameter meter that uses the HI1230B pH electrode and HI7669AW temperature probe (both included) using separate connections.
Manual calibration is performed at one or two points by adjusting the trimmers on the front panel. Capable of measuring pH/mV and °C, this meter is great for field work, providing one meter for multiple uses. This instrument is ideal for applications that require a custom calibration point. Manual calibration can be extremely useful in order to achieve better accuracy.
  • Automatic Temperature Compensation (ATC)
  • Two-point Calibration
  • Compact, heavy-duty, and waterproof protected casing
  • Temperature probe and pH electrode included
  • Can also be used for ORP measurements with the HI3131B probe (sold separately)
Specifications
Includes pH electrode, temperature probe, 1m cable, pH 4.01 / 7.01 buffer solution sachets, electrode cleaning solution sachets, calibration screwdriver, battery, protective case, instructions
Accuracy (Temperature) ±0.4°C
Accuracy (mV) ±1
Accuracy (pH) ±0.01
For Use With (Application) Ideal for education and field applications
Calibration Manual via Trimmers
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) ±1999
Resolution (mV) 1mV
Calibration Points 2
Temperature Range (Metric) 0°C to 100°C
Temperature Range (English) 32°F to 212°F
Resolution (Temperature) 0.1°C
Temperature Probe Calibration Slope/Offset Calibration: from 80 to 110% ±1pH
Inputs BNC connectors
Dimensions (L x W x H) 145 x 80 x 36 mm
Weight (Metric) 230 g
Weight (English) 8.1 oz.
Material Stainless Steel Temperature Probe, PEI-body pH Electrode,
Probe Type HI1230B pH Electrode and HI7669AW Temperature Probe, with BNC connector
Detection Range 0 to 14 pH
Temperature Compensation Automatic, 0 to 70°C
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 0°C to 50°C
Battery Type 9V
Type Analog pH/mV/Temp Meter
Battery Life 100 hr.