Learn More
Brady™ Porta-Pack™ Write-on Self-laminating Marker
Product Code. 11734654 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
39.7 mm
57.15 mm
Unit Size:
1 pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
11734654 57.15 mm 1 pack
10179092 39.7 mm 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11734654 Supplier Brady™ Supplier No. 035403
  • ONLINE EXCLUSIVE PRICE
    47.00€
    39.74€ / Pack of 1
    Save 7.26€
    15% Off
Estimated Shipment: 28-12-2026
Three different Porta-Pack™ Write-on Self-laminating Marker sizes are available to suit all of your labelling needs.
  • Porta-Pack™ books are a convenient way to ensure the wire markers you need are always on hand.
  • Transparent portion of label over-wraps provides a snug fitting, self-laminating shield that resists oil, water and solvents.
  • Labels can be used for applications above or below ground.
  • Three different marker sizes are available to suit all of your labelling needs.
Specifications
Certifications/Compliance ASTM D 1000, ASTM D 2979
Description Porta-Pack Wire Marker Books - Write-on Self-laminating Markers
Adhesive Type Acrylic, Permanent
Color White, Transparent
Finish Matte
Format Label
Material Self-laminating Vinyl
Print Technology Dot Matrix
Product Type Wire and Cable Markers
Sterility Non-sterile
For Use With (Application) Wire and cable labelling
Includes 30 Blank Write-On Markers (1 x 3 in.);15 Blank Write-On Markers (1 x 6 in.)
Min. Operating Temperature (Metric) -40°C
No. per Pack 45 Labels/Pk.
Quantity 45 Pk.
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 57.15 x 25.4 mm
Length (English) 2.25 in.
Width (English) 1 in.
Width (Metric) 25.4 mm
Width (English) Printable Area 1 in.
Width (Metric) Printable Area 25.40 mm
Height (English) 2.25 in.
Height (English) Printable Area 1 in.
Height (Metric) 57.15 mm
Height (Metric) Printable Area 25.40 mm
Diameter (Metric) 40 mm
Alternate Part Number PWC-PK-36