Learn More
ROBU VitraPOR™ Porous Glass Beads
Product Code. 16813306
Change view
Click to view available options
Diameter (Metric):
1.5 mm
8 mm
Unit Size:
1 L
Product Code. Diameter (Metric) unitSize
16813306 1.5 mm 1 L
16823306 8 mm 1 L
2 options
Product Code. 16813306
Brand: ROBU VitraPOR™ 80080
Substrate for cell cultivation.
  • Pure, binder-free USP-class I borosilicate glass 3.3
  • The special process uses no corrosives and produces a porous structure with smooth, yet large surface areas
  • The glass is completely bio-compatible and its high electrical surface charge (Zeta-Potential) helps to stimulate cell growth
Specifications
Material Borosilicate Glass (3.3)
Diameter (Metric) 1.5 mm
Description Pore Size: 2
Quantity 1 Ea.