missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Porcine TGF-beta 1 ELISA Kit (Chemiluminescence)

Product Code. 30279685 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30279685

Brand: Novus Biologicals™ NBP343240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemiluminescence ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 13.7 to 10000 pg/mL
Assay Sensitivity 5.7 pg/mL
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Sample Type Serum, Plasma, Tissue Homogenates, Cell Lysates, Cell Culture Supernatant
Target Species Porcine
For Use With (Application) ELISA
Gene ID (Entrez) 7040
Gene Symbol TGFB1
Kit Contents Pre-coated 96T Strip Plate, Plate sealer for 96 wells, Standard, Standard Diluent, Detection Reagent A, Assay Diluent A, Detection Reagent B, Assay Diluent B, TMB Substrate, Stop Solution, Wash Buffer (30X concentrate), Instruction manual
Quantity 1 Kit
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.