Learn More
R&D Systems Porcine Luminex™ Discovery Assay
Product Code. 18732164
Click to view available options
Quantity:
1 Plex
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
3 Plex
4 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Product Code. 18732164
Brand: R&D Systems LXSAPM10
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 Plex
Species Porcine
Binding Target Multiplex Cytokine Assay Kits
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.