missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Porcine IL-1ra/IL-1F3 DuoSet ELISA
Description
The Porcine IL-1ra/IL-1F3 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies porcine IL-1ra/IL-1F3/IL1RN.
Specifications
Specifications
| Product Type | Solid Phase Sandwich ELISA |
| Species | Porcine |
| Binding Target | IL-1ra/IL-1F3/IL1RN |
| Assay Range | 93.80 to 6000 pg/mL |
| Format | 96-well Microplate |
| Gene ID (Entrez) | 3557 (Human); 16181 (Mouse); 60582 (Rat); 397499 (Porcine) |
| Sample Type | Cell Culture Supernatants, Serum, EDTA Plasma, Heparin Plasma |
| Kit Contents | 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?