Learn More
Haldenwanger™ Porcelain Spouted Mortar
Product Code. 12363328
Change view
Click to view available options
Diameter (Metric) Outer:
65mm
130mm
150mm
180mm
250mm
330mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
12363328 150mm Each
12393328 330mm Each
12373328 180mm Each
12383328 250mm Each
10720603 130mm Each
10403925 65mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12363328 Supplier Haldenwanger™ Supplier No. L 55/6A/RAUH
  • ONLINE EXCLUSIVE PRICE
    86.05€
    76.43€ / Pack of 1
    Save 9.62€
    11% Off
Estimated Shipment: 15-12-2026
Use these mortars for a variety of laboratory applications. Haldenwanger™ Porcelain Spouted Mortars are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • With spout for easy pouring
  • Complies with DIN VDE 0335, type C 100
Specifications
Diameter (Metric) Outer 150mm
Product Type Mortar
Material Porcelain
Height (Metric) 70mm
Includes With spout
Capacity (Metric) 650 mL