Learn More
Heinz Herenz Porcelain Pestle with Rough Finish
Product Code. p-200111935
Click to view available options
Capacity (Metric):
54 mm
115 mm
135 mm
150 mm
175 mm
210 mm
250 mm
280 mm
Unit Size:
Each
Product Code. 30158514
Brand: Heinz Herenz 1071081
Porcelain pestle with rough finish.
  • Rough finish
  • Porcelain
Specifications
Capacity (Metric) 210 mm
Finish Rough
Autoclavable Autoclavable
Product Type Pestle
Material Porcelain
Type Pestle