Learn More
Haldenwanger™ Porcelain Pestles
Product Code. 10405011
Change view
Click to view available options
Length (Metric):
115mm
150mm
175mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10405011 115mm Each
10693291 150mm Each
11930897 175mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10405011 Supplier Haldenwanger™ Supplier No. L 56/00/RAUH
  • ONLINE EXCLUSIVE PRICE
    16.35€
    12.76€ / Pack of 1
    Save 3.59€
    21% Off
Estimated Shipment: 10-12-2026
Use these porcelain pestles for a wide range of applications. Haldenwanger™ Porcelain Pestles are available glazed or unglazed with head sizes from 24 to 85mm in diameter. Stringent inspections, starting with raw materials and continuing down to finished products, guarantee constant quality.
  • Grinding surfaces are unglazed
  • Excellent chemical resistance
  • Laboratory porcelain complies with DIN EN 60672-3,Group C100, Type C110
Specifications
Diameter (Metric) Outer 24mm
Product Type Pestle
Material Porcelain
Length (Metric) 115mm
Finish Unglazed