Learn More
Haldenwanger™ Porcelain Pestle
Product Code. 11940897
Change view
Click to view available options
Diameter (Metric) Outer:
60mm
180mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
11940897 180mm Each
12383338 60mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11940897 Supplier Haldenwanger™ Supplier No. L 56/4/RAUH
  • ONLINE EXCLUSIVE PRICE
    39.60€
    35.17€ / Pack of 1
    Save 4.43€
    11% Off
Estimated Shipment: 23-11-2026
Use these pestles for a variety of laboratory applications. Haldenwanger™ Porcelain Pestles are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • Grinding surfaces are unglazed
  • Complies with DIN VDE 0335, type C 100
Specifications
Diameter (Metric) Outer 180mm
Product Type Pestle
Material Porcelain