Learn More
Porcelaines Avignon™ High Form Porcelain Crucibles
Click to view available options
Capacity (Metric):
6 mL
14 mL
17 mL
30 mL
80 mL
120 mL
250 mL
Unit Size:
Pack of 6
Description
- High walls
- Withstand high temperatures and rapid temperature changes
- Porcelain
Specifications
Specifications
| Material | Porcelain |
| Capacity (Metric) | 14 mL |
| Diameter (Metric) Outer | 30mm |
| Height (Metric) | 33mm |
| Product Type | Crucible |