Learn More
Porcelaines Avignon™ Porcelain Gooch Crucibles
Product Code. 11708463
Click to view available options
Diameter (Metric) Outer:
35 mm
Unit Size:
Pack of 6
Product Code. 11708463
Brand: Porcelaines Avignon™ 11403
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Order Info
DiameterMetricOuter
Specifications
Material Porcelain
Diameter (Metric) Outer 35 mm
Height (Metric) 40mm
Product Type Gooch crucible