Learn More
Haldenwanger™ Porcelain Crucible Lids
Product Code. 10557301
Click to view available options
Diameter (Metric) Outer:
30mm
40mm
45mm
60mm
75mm
87mm
92mm
Unit Size:
Each
Pack of 5
Product Code. 10557301
Brand: Haldenwanger™ L 79D/8
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use these lids with porcelain crucibles. Haldenwanger™ Porcelain Crucible Lids are constructed from optimized laboratory porcelain.
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Material Porcelain
Diameter (Metric) Outer 30mm
Product Type Crucible Lid