Learn More
Avignon Ceramic Porcelain Buchner Funnel
Product Code. 11708413
Click to view available options
Material:
Porcelain
Unit Size:
Each
Product Code. 11708413
Brand: Avignon Ceramic 12706
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Porcelain Buchner Funnel is designed for rapid filtration and can withstand high temperatures.
  • Liquid and gas tight
  • Chemically neutral
  • High resistance to thermal and physical shocks
Specifications
Diameter (Metric)Top 80 mm
Material Porcelain
Height (Metric) 135 mm
Max. Temperature (Metric) 1100°C