Learn More
Rogo-Sampaic Porcelain Buchner Funnels
Product Code. 11797335
Change view
Click to view available options
Capacity (Metric):
40 mL
80 mL
150 mL
265 mL
490 mL
660 mL
1080 mL
2000 mL
4200 mL
10 L
Diameter (Metric) Plate:
45mm
55mm
70mm
90mm
110mm
125mm
150mm
185mm
240mm
320mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Plate unitSize
11797335 2000 mL 185mm Each
11707335 150 mL 70mm Each
11797325 80 mL 55mm Each
11787325 40 mL 45mm Each
11717335 265 mL 90mm Each
11707345 4200 mL 240mm Each
11737335 660 mL 125mm Each
11747335 1080 mL 150mm Each
11727335 490 mL 110mm Each
11717345 10 L 320mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 11797335 Supplier Rogo-Sampaic Supplier No. PNF015
Only null left
Add to basket
Add to basket
Excellent for separating liquids from solids in the laboratory. Porcelain Buchner Funnels provide a quick and easy pressure assisted filtration method.
  • White porcelain
  • Available for a range of volumes and filter diameters
  • Applications include collecting recrystalized compounds
  • Excellent thermal and chemical resistance
Specifications
Capacity (Metric) 2000 mL
Material Porcelain
Diameter (Metric) Plate 185mm
Diameter (Metric) Disc 185mm
Length (Metric) Stem 117mm
Quantity 1Each
Product Type Buchner Funnel