Learn More
VITLAB™ POM Blunt Tip Forceps
Product Code. 11857910
Click to view available options
Length (Metric):
180 mm
250 mm
Unit Size:
Pack of 25
Pack of 5
Product Code. 11857910
Brand: VITLAB™ 68399
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Grasp and hold small objects easily and securely. VITLAB™ POM Blunt Tip Forceps features a rounded tip and grooved body for optimal grip. Constructed of polyoxymethylene for high elasticity, durability and chemical resistance. Packs of five forceps in in two sizes are available.
Non-locking thumb forceps are designed to grasp and hold small objects where fingers cannot reach or when a higher level of grip stability or sterile handling is required. These yellow polyoxymethylene tweezers are designed for optimal object handling.
  • Blunt, rounded ends ensure secure grasping of small objects
  • Grooves on the outside provide optimum grip for secure handling
  • Long forceps (180 and 250mm) provide good reach for use with blotting and decanting
  • Constructed of durable polyoxymethylene (POM)
    • Provides high elasticity for easy operation
    • Can be used with aliphatic alcohols, ether, aldehydes, hydrocarbons, ketones and alkalis
    • Not recommended for use with ester, acids and oxidizing agents or in a microwave
  • Can be autoclaved for cleaning/sterilization (frequent autoclaving may reduce mechanical stability)
Specifications
Tip Style Blunt
No. per Pack 5/Pack
Material POM
Length (Metric) 250 mm
Product Type Forceps, Non-locking Thumb
Finish Yellow
For Use With (Application) Bottling and decanting
Style Grooves on the outside