Learn More
Navailles Polyurethane Asynchronous Chair
Product Code. 17426182
Change view
Click to view available options
Seating Height:
460 to 590 mm
560 to 800 mm
Wheel Type:
None
Wheels with rubber tires
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Seating Height Wheel Type Unit Size
17426182 560 to 800 mm None Each
17416182 460 to 590 mm Wheels with rubber tires Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17426182 Supplier Navailles Supplier No. 7NG 40GHLP 01 905
  • ONLINE EXCLUSIVE PRICE
    522.22€
    452.00€ / Pack of 1
    Save 70.22€
    13% Off
View all promotions
Estimated Shipment: 26-11-2026
The Polyurethane Asynchronous Chair is fully and easily adjustable, featuring a comfortable and airy surface. Constructed without staples nor seams.
  • 5-branch, scuff-resistant, reinforced chrome base
  • Chrome gas-lift column for height adjustment with shock absorber
  • Ergonomic seat and backrest in black soft polyurethane
  • Independently adjustable seat and backrest tilt with one fingertip lever
Especificaciones
Color Black
Adjustable Yes
Arm Style No Armrests
Certifications/Compliance EN 1021-1, EN 1021-2
For Use With (Application) Laboratories, Chemical Industry
Lift Type Gas-lift column
Load Bearing Capacity (English) 265 lb.
Load Bearing Capacity (Metric) 120 kg
Seating Height 560 to 800 mm
Warranty 8 Years
Weight (English) 30.9 lb.
Weight (Metric) 14.1 kg
Wheel Type None
Width (English) 23.2 in.
Width (Metric) 59 cm
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.