Learn More
Bel-Art™ SP Scienceware™ Polytetrafluoroethylene Spinbar™ Flexible Magnetic Retrivers
Product Code. 10603095
Change view
Click to view available options
Length (Metric):
33 mm
365 mm
Color:
Blue
Red
White
Yellow
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Color unitSize
10603095 33 mm White Each
11765064 33 mm Red Each
11729695 33 mm Yellow Each
11775064 33 mm Blue Each
12301288 365 mm Red Each
12311288 365 mm Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10603095 Supplier Bel-Art™ Supplier No. H377740101
Flexible magnetic Stirbar retriever recovers stir bars from lab sink drains and containers.
  • Flexible, Teflon™ rod with powerful Alnico Teflon™ coated magnet
  • Available in four colors, two magnet sizes
  • Flexible white rod is 33cm (13 in.) long
  • Autoclavable
Specifications
Color White
Length (Metric) 33 mm
Material PTFE