Learn More
Saint-Gobain Chemware™ PTFE Cord
Product Code. 10052664
Click to view available options
Diameter (Metric):
0.8 mm
Unit Size:
61m
Product Code. 10052664
Brand: Saint-Gobain D1069137
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Saint Gobain Performance Plastics™ Chemware™ PTFE Cord resists adhesion of practically any material and can be used to secure the edges of laboratory matting to create a dipping basket for corrosive baths.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.8 mm
Length (Metric) 61 m (per Roll)
Material PTFE