Learn More
Buerkle™ Polystyrene Volumetric Spoons
Product Code. 13427888 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0.5 mL
1 mL
Unit Size:
Pack of 100
Product Code. 13427888
Brand: Buerkle™ 53340005
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
The capacity is embossed into the spoon handle. Available as a set with 8 volumetric spoons or separate in bags of 100 volumetric spoons.
  • Polystyrene, blue
  • Round shape, easy emptying
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 0.5 mL
Color Blue
Product Type Volumetric Spoon
Material Polystyrene