Learn More
Semadeni™ Polystyrene Tweezers
Click to view available options
Length (Metric):
120 mm
125 mm
Style:
Bulk packed
Individually wrapped
Unit Size:
Pack of 100
Description
- Transparent polystyrene (PS) general-purpose tweezers
Specifications
Specifications
| Tip Style | Straight, Round |
| Material | Polystyrene |
| Length (Metric) | 120 mm |
| Product Type | Tweezers |
| Quantity (No. per Case) | 100/Pack |
| Type | Tweezers |
| Style | Individually wrapped |