Learn More
Multiroir™ Polystyrene Rectangular Trays
Product Code. p-8003230
Click to view available options
Height (Metric):
34 mm (Inner), 37 mm (Outer)
40 mm (Inner), 42 mm (Outer)
42 mm
Color:
Blue
Green
Red
White
Yellow
Unit Size:
Each
Product Code. 11778361
Brand: Multiroir™ 39105
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Organize your workspace with these durable trays. Multiroir™ Polystyrene Rectangular Trays are widely used in the laboratory and available in a variety of colors and styles.
  • Multi-purpose
  • Polystyrene
  • Rectangular
Specifications
Color Blue
Height (Metric) 34 mm (Inner), 37 mm (Outer)
Length (Metric) 175 mm (Inner), 205 mm (Outer)
Material Polystyrene
Type Tray
Width (Metric) 100 mm (Inner), 130 mm (Outer)
No. per Pack 1 Pack
Shape Rectangular