Description
- Dishes work well in robotic dispensers due to flatness and uniform height.
- Models 14007-10 and -11 are square in shape.
- Models 14007-03 and -05 are 5 mm deeper and models 14007-04 and -14 are 10 mm deeper than standard dish depths to accommodate longer culture periods.
- Sterilized by gamma irradiation.
Specifications
Specifications
| Color | Transparent |
| Disposable | Single-use |
| Material | Polystyrene |
| Shape | Square |
| Sterility | Sterile |
| Type | Petri Dish |
| For Use With (Application) | For culturing fungi, bacteria, and other microorganisms |
| Quantity | 300 Pk. |
| Height (Metric) | 25 mm |
| Height (English) | 1 in. |
| Show More |