Learn More
Greiner Bio-One Polystyrene Container for Plant Tissue Culture
Description
- Maximum light transmission, rapid, successful growth
- Used as transport containers
Service Plan
No
Warranty
No
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Round |
| Color | Clear |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Sterile |
| Type | Culture Container |
| For Use With (Application) | Plant tissue culture |
| Show More |