Learn More
Greiner Bio-One Polystyrene CELLSTAR Cell Culture Tubes
Product Code. 10199900
Click to view available options
Closure Type:
Dual-position Vent Stopper
Screw Cap
Capacity (Metric):
12 mL
14 mL
Unit Size:
Pack of 1000
Pack of 750
Product Code. 10199900
Brand: Greiner Bio-One 163160
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Improved cell adhesion due to physical surface treatment.
  • TC treated
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Red
Diameter (Metric) 17 mm
Product Type Culture Tube
Closure Type Screw Cap
Length (Metric) 100 mm
Material Polystyrene
Surface Treatment TC Treated
Sterility Sterile
Capacity (Metric) 12 mL
Closure Included Yes