Learn More
Kartell™ Polystyrene Biological Specimen Containers
Manufactured with crystal clear polystyrene for optimal visibility. Kartell™ Polystyrene Biological Specimen Containerrs are available with polyethylene and polypropylene color coded screw caps.
155.00€ - 180.00€
Specifications
Product Type | Biological Specimen Container |
---|---|
Closure Included | Yes |
Material | Polystyrene |
Sterility | Sterile |
Certifications/Compliance | CE and IVD |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
10102381
|
Kartell™
0269600 |
200 mL | 66 mm | 86 mm |
155.00€
Pack of 200 |
In Stock
Log in to see stock. |
|||
10355631
|
Kartell™
0563100 |
60 mL | 39 mm | 65 mm |
180.00€
Pack of 400 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||
Description
- Sterile containers are individually wrapped
- CE marked;IVD approved
Specifications
Biological Specimen Container | |
Polystyrene | |
CE and IVD | |
Polypropylene | |
Urine and biological specimen collection | |
Yes |
Yes | |
Sterile | |
Red | |
Screw Cap | |
Polypropylene Screw Cap |