Learn More
Polysorbate 80, Multi-Compendial, N.F., J.T. Baker™
Product Code. 15469933 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
20 mL
200 mL
1 L
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
Steel Drum
Unit Size:
1 L
19 L
200mL
20mL
4 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15469933 20 mL Narrow Mouth Amber Glass Bottle 20mL
10554683 200 mL Narrow Mouth Amber Glass Bottle 200mL
15528894 1 L Narrow Mouth Amber Glass Bottle 1 L
12615457 1 L Narrow Mouth Amber Glass Bottle, w/ Septum 1 L
15362724 4 L Narrow Mouth Amber Glass Bottle 4 L
15538894 19 L Steel Drum 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15469933 Supplier Avantor J.T.Baker Supplier No. 411702
  • ONLINE EXCLUSIVE PRICE
    193.83€
    157.62€ / 20mL
    Save 36.21€
    18% Off
 Restricted Item
In Stock
Add to basket
Edge
Chemical Identifiers
CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
Specifications
pH 5.5
Quantity 20 mL
Flash Point 148.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.08kg
Grade Multi-Compendial/NF
Chemical Name or Material Polysorbate 80