Learn More
Epredia™ Polysine Adhesion Slides
Product Code. 10219280 Shop All Epredia Products
Change view
Click to view available options
Dimensions (L x W):
75 x 25 mm
Unit Size:
Pack of 72
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10219280 75 x 25 mm Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10219280 Supplier Epredia™ Supplier No. J2800AMNZ
  • ONLINE EXCLUSIVE PRICE
    33.43€
    17.76€ / Pack of 72
    Save 15.67€
    46% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Epredia™ Polysine adhesion slides are electrostatically and biochemically adhesive. The preparation is first adhered by electrostatic attraction and then fixed in place by biochemical binding. Polysine adhesion slides are best suited to paraffin-embedded sue slices from human sources fixed in formalin, alcohol or Bouin solution. For tissue slices 2 to 5 micrometers thick.
  • No time wasted on slide-coating proceduress
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 75 mm
Width (Metric) 25 mm
Thickness (Metric) 1 mm
Color White
For Use With (Application) Tissue slices 2 to 5μm thick
Dimensions (L x W) 75 x 25 mm