Learn More
InterLab™ Supply PolySeed™ Microbiol BOD Capsules
Product Code. 15804448 Shop All Cole-Parmer Products
Change view
Click to view available options
Quantity:
50 capsules per bottle
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15804448 50 capsules per bottle Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15804448 Supplier InterLab™ Supply Supplier No. 5320033
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
PolySeed™ Microbiol BOD Capsules - Easy to use capsules contain a special blend of microbial cultures that provide a uniform standard for BOD analysis.
  • Fresh consistent seeds each time
  • One capsule performs approximately 100 tests
  • Easy to use
Specifications
For Use With (Application) BOD analysis
No. of Tests One capsule performs 100 tests
Quantity 50 capsules per bottle
Type BOD microbial BOD capsules