Learn More
Polypyrrole (undoped, extent of labeling: ~20 wt. % loading, composite with carbon black), TRC
Specifications
Specifications
| Chemical Name or Material | Polypyrrole |
| CAS | 30604-81-0 |
| Molecular Formula | (C4 H5 N)mon |
| Quantity | 500 mg |
| Synonym | Basotronic PYR, Coreron YE, Lutamer ES 9567, Lutamer P 160, PPY 12, Poly(1H-pyrrole), Polyazole, Pyrrole Homopolymer, Pyrrole Polymer, ST-Poly |
| InChI Formula | InChI=1S/C4H5N/c1-2-4-5-3-1/h1-5H |
| SMILES | C1=CC=CN1 |
| Molecular Weight (g/mol) | 67.089 |
| Formula Weight | 67.042 |